Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMOD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113399
Description
TMOD3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.
Specifications
| TMOD3 | |
| Polyclonal | |
| Unconjugated | |
| Tmod3 | |
| actin filament pointed-end capping protein; Tmod3; tropomodulin 3; tropomodulin 3 (ubiquitous); tropomodulin 3; ubiquitous tropomodulin; tropomodulin-3; Tropomodulin-3-like protein; Ubiquitous tropomodulin; ubiquitous tropomodulin U-Tmod; Unknown (protein for MGC:127550); UTMOD; U-Tmod | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29766 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NYL9 | |
| Tmod3 | |
| Recombinant Human Tropomodulin-3 protein (1-352AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction