Learn More
TMOD3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305451AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.Specifications
TMOD3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NYL9 | |
TMOD3 | |
Between 150 and 200 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TMOD3 | |
UTMOD, tropomodulin-3, ubiquitous tropomodulin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29766 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.