Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMOD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110500 Supplier Invitrogen™ Supplier No. PA5110500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSYQKELEK YRDIDEDEIL RTLSPEELEQ LDCELQEMDP ENMLLPAGLR QRDQTKKSPT GPLDREALLQ YLEQQALEVK ERDDLVPFTG EKKGKPYIQP KREIPAEEQI TLEPELEEAL AHATDAEMCD IAAILDMYTL MSNKQYYDAL CSGEICNTEG ISSVVQPDKY KPVPDEPPNP TNIEEILKRV RSNDKELEEV NLNNIQDIPI PMLSELCEAM KANTYVRSFS LVATRSGDPI ANAVADMLRE NRSLQSLNIE SNFISSTGLM AVLKAVRENA TLTELRVDNQ RQWPGDAVEM EMATVLEQCP SIVRFGYHFT QQGPRARAAQ AMTRNNELRR QQKKR.

TMOD4 blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMOD4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMOD4
Gene Accession No. Q9JLH8, Q9NZQ9
Gene Alias actin-capping protein; MTMOD; skeletal muscle tropomodulin; skeletal tropomodulin; SK-TMOD; TMOD4; tropomodulin 4; tropomodulin 4 (muscle); tropomodulin-4
Gene Symbols TMOD4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-345 of human TMOD4 (NP_037485.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295261, 29765, 50874
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.