Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tmp21/p23 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11057587AF405
Description
Tmp21/p23 Polyclonal specifically detects Tmp21/p23 in Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Tmp21/p23 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated | |
P49755 | |
TMED10 | |
A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 50-150). [Swiss-Prot# P49755] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus species | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein | |
Rabbit | |
21 kDa | |
0.1 mL | |
Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction | |
10972 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction