Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tmp21/p23 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11057587G
Description
Tmp21/p23 Polyclonal specifically detects Tmp21/p23 in Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Tmp21/p23 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated | |
P49755 | |
TMED10 | |
A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 50-150). [Swiss-Prot# P49755] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus species | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein | |
Rabbit | |
21 kDa | |
0.1 mL | |
Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction | |
10972 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction