Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMPRSS11A Polyclonal Antibody
TMPRSS11A Polyclonal Antibody
Supplier: Thermo Scientific PA572715
Description
TMPRSS11A Polyclonal Antibody for Western Blot, ELISA

Specifications
TMPRSS11A | |
Polyclonal | |
Unconjugated | |
TMPRSS11A | |
airway trypsin-like protease 1; Desc3; DESC-3; ECRG1; epidermal type II transmembrane serine protease; epidermal type-II transmembrane serine protease; Esophageal cancer-susceptibility gene 1 protein; Gm7; Hatl1; HESP; Serine protease DESC3; TMPRSS11A; Transmembrane protease serine 11A; transmembrane protease, serine 11A | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
194597, 339967, 686581 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q3UQ41, Q6ZMR5 | |
TMPRSS11A | |
ECRG1 antibody for a 16 amino acid synthetic peptide near the carboxy terminus of human ECRG1. The immunogen is within amino acids 300 - 350 of ECRG1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction