Learn More
Invitrogen™ TMPRSS11A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597130
Description
Immunogen sequence: SASFQPNLTV HITGFGALYY GGESQNDLRE ARVKIISDDV CKQPQVYGND IKPGMFCAGY MEGIYDACRG DSGGPLVTRD LKDTWYLIGI VSWGDNCGQK DKPGVYTQVT YYRNWIASKT GI; Positive Samples: HeLa, A-549, SW620, Mouse liver, Rat liver; Cellular Location: Membrane, Single-pass type II membrane protein.
Probable serine protease which may play a role in cellular senescence. Overexpression inhibits cell growth and induce G1 cell cycle arrest.
Specifications
TMPRSS11A | |
Polyclonal | |
Unconjugated | |
TMPRSS11A | |
airway trypsin-like protease 1; Desc3; DESC-3; ECRG1; epidermal type II transmembrane serine protease; epidermal type-II transmembrane serine protease; Esophageal cancer-susceptibility gene 1 protein; Gm7; Hatl1; HESP; Serine protease DESC3; TMPRSS11A; Transmembrane protease serine 11A; transmembrane protease, serine 11A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
194597, 339967, 686581 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.08 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UQ41, Q6ZMR5 | |
TMPRSS11A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 300-421 of human TMPRSS11A (NP_8724123). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.