Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMPRSS11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120253 Supplier Invitrogen™ Supplier No. PA5120253
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, PC-12. The target is usually found in the following locations: Membrane, Single-pass type II membrane protein.

Probable serine protease which may play a role in cellular senescence. Overexpression inhibits cell growth and induce G1 cell cycle arrest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS11A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMPRSS11A
Gene Accession No. Q3UQ41, Q6ZMR5
Gene Alias airway trypsin-like protease 1; Desc3; DESC-3; ECRG1; epidermal type II transmembrane serine protease; epidermal type-II transmembrane serine protease; Esophageal cancer-susceptibility gene 1 protein; Gm7; Hatl1; HESP; Serine protease DESC3; TMPRSS11A; Transmembrane protease serine 11A; transmembrane protease, serine 11A
Gene Symbols TMPRSS11A
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 194597, 339967, 686581
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.