Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMPRSS11D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587660 Supplier Invitrogen™ Supplier No. PA587660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFDQKSYFYR SSFQLLNVEY NSQLNSPATQ EYRTLSGRIE SLITKTFKES NLRNQFIRAH VAKLRQDGSG VRADVVMKFQ FTRNNNGASM KSRIESVLRQ MLNNSGNLEI NPSTEITSLT DQAAANWLIN ECGAGPDLIT LSEQRILGGT EAEEGSWPWQ VSLRLNNAHH CGGSLINNMW ILTAAHCFRS NSNPRDWIAT SGISTTFPKL RMRVRNILIH NNYKSATHEN DIALVRLENS VTFTKDIHSV C; Positive Samples: HepG2, Mouse liver, Mouse kidney, Mouse skeletal muscle, Rat liver, Rat kidney; Cellular Location: Cell membrane, Single-pass type II membrane protein.

The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS11D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMPRSS11D
Gene Accession No. O60235, Q8VHJ4, Q8VHK8
Gene Alias Adrenal secretory serine protease; airway spesific trypsin-like protease; airway trypsin like protease; airway trypsin-like protease; AsP; AST; AT; BC020151; HAT; Mat; MGC150587; MGC150588; Rat; TMPRSS11D; Transmembrane protease serine 11D; Transmembrane protease serine 11D catalytic chain; Transmembrane protease serine 11D non-catalytic chain; transmembrane protease, serine 11D; transmembrane serine protease 11D
Gene Symbols TMPRSS11D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 42-292 of human TMPRSS11D (NP_0042531).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231382, 64565, 9407
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.