Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMPRSS11D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530927 Supplier Invitrogen™ Supplier No. PA530927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS11D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TMPRSS11D
Gene Accession No. O60235
Gene Alias Adrenal secretory serine protease; airway spesific trypsin-like protease; airway trypsin like protease; airway trypsin-like protease; AsP; AST; AT; BC020151; HAT; Mat; MGC150587; MGC150588; Rat; TMPRSS11D; Transmembrane protease serine 11D; Transmembrane protease serine 11D catalytic chain; Transmembrane protease serine 11D non-catalytic chain; transmembrane protease, serine 11D; transmembrane serine protease 11D
Gene Symbols TMPRSS11D
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human TMPRSS11D. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9407
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.