Learn More
Invitrogen™ TMPRSS11E Polyclonal Antibody

Description
The antibody detects endogenous levels of total TMPRSS11E protein.
Specifications
Specifications
| Antigen | TMPRSS11E |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | TMPRSS11E |
| Gene Accession No. | Q9UL52 |
| Gene Alias | DESC1; Serine protease DESC1; TMPRSS11E; TMPRSS11E2; transmembrane protease serine 11E; Transmembrane protease serine 11E catalytic chain; Transmembrane protease serine 11E non-catalytic chain; Transmembrane protease serine 11E2; transmembrane protease, serine 11E; transmembrane protease, serine 11E2; UNQ742/PRO1461 |
| Gene Symbols | TMPRSS11E |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.