Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMPRSS11E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562306 Supplier Invitrogen™ Supplier No. PA562306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLAHMLLICR FHSTEDPETV DKIVQLVLHE KLQDAVGPPK VDPHSVKIKK INKTETDSYL NHCCGTRRSK TLGQSLRIVG GTEVEEGEWP Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.

Serine protease which possesses both gelatinolytic and caseinolytic activities. Shows a preference for Arg in the P1 position.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS11E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMPRSS11E
Gene Accession No. Q9UL52
Gene Alias DESC1; Serine protease DESC1; TMPRSS11E; TMPRSS11E2; transmembrane protease serine 11E; Transmembrane protease serine 11E catalytic chain; Transmembrane protease serine 11E non-catalytic chain; Transmembrane protease serine 11E2; transmembrane protease, serine 11E; transmembrane protease, serine 11E2; UNQ742/PRO1461
Gene Symbols TMPRSS11E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMPRSS11E. Recombinant protein control fragment (Product #RP-99514).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.