Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMPRSS11F Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TMPRSS11F |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TMPRSS11F Polyclonal specifically detects TMPRSS11F in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TMPRSS11F | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
389208 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NKDPTQWIATFGATITPPAVKRNVRKIILHENYHRETNENDIALVQLSTGVEFSNIVQRVCLPDSSIKLPPKTSVFVTGFGSIVDDGPIQNTLRQARVETISTDVCNRKDVYDG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ16046, transmembrane protease serine 11F, transmembrane protease, serine 11F | |
TMPRSS11F | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title