Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMPRSS12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570159
Description
This target displays homology in the following species: Guinea Pig: 86%; Human: 100%; Rabbit: 79%; Rat: 86%.
TMPRSS12 (transmembrane protease serine 12) is a 348 amino acid single-pass membrane protein that belong to the peptidase S1 family and contains one peptidase S1 domain. The gene that encodes TMPRSS12 consists of nearly 45,000 bases and maps to human chromosome 12q13.12. Encoding over 1,100 genes within 132 million bases, chromosome 12 makes up about 4.5% of the human genome. A number of skeletal deformities are linked to chromosome 12, including hypochondrogenesis, achondrogenesis and Kniest dysplasia. Noonan syndrome, which includes heart and facial developmental defects among the primary symptoms, is caused by a mutant form of PTPN11 gene product, SH-PTP2. Chromosome 12 is also home to a homeobox gene cluster, which encodes crucial transcription factors for morphogenesis, and the natural killer complex gene cluster, encoding C-type lectin proteins which mediate the NK cell response to MHC I interaction. Trisomy 12p leads to facial development defects, seizure disorders and a host of other symptoms which vary in severity depending on the extent of mosaicism.
Specifications
TMPRSS12 | |
Polyclonal | |
Unconjugated | |
Tmprss12 | |
CT151; rCG50645-like; RGD1560236; testicular tissue protein Li 208; TMPRSS12; transmembrane (C-terminal) protease, serine 12; transmembrane protease serine 12; transmembrane protease, serine 12; transmembrane serine protease 12 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
283471 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86WS5 | |
Tmprss12 | |
synthetic peptide directed towards the middle region of human TMPRSS12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction