Learn More
Invitrogen™ TMPRSS13 Polyclonal Antibody

Description
TMPRSS13 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TMPRSS13 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TMPRSS13 |
| Gene Accession No. | Q9BYE2 |
| Gene Alias | membrane-type mosaic serine protease; Mosaic serine protease; MSP; MSPL; MSPS; TMPRSS11; TMPRSS13; Transmembrane protease serine 13; transmembrane protease, serine 11; transmembrane protease, serine 13 |
| Gene Symbols | TMPRSS13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.