Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMPRSS15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144219 Supplier Invitrogen™ Supplier No. PA5144219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMPRSS15 Polyclonal Antibody for IHC (P), IP, ELISA

This gene encodes an enzyme that converts the pancreatic proenzyme trypsinogen to trypsin, which activates other proenzymes including chymotrypsinogen and procarboxypeptidases. The precursor protein is cleaved into two chains that form a heterodimer linked by a disulfide bond. This protein is a member of the trypsin family of peptidases. Mutations in this gene cause enterokinase deficiency, a malabsorption disorder characterized by diarrhea and failure to thrive. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS15
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMPRSS15
Gene Accession No. P98073
Gene Alias enterokinase; enterokinase catalytic subunit; Enterokinase light chain; enteropeptidase; Enteropeptidase catalytic light chain; Enteropeptidase non-catalytic heavy chain; Entk; protease, serine, 7 (enterokinase); PRSS7; Serine protease 7; Tmprss15; Transmembrane protease serine 15; transmembrane protease, serine 15
Gene Symbols TMPRSS15
Host Species Rabbit
Immunogen Recombinant Human Enteropeptidase protein (40-300AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5651
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.