Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMPRSS2 Antibody (1038105), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB107231PECY7
Description
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human samples. It is validated for Flow CytometrySpecifications
TMPRSS2 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1038105 | |
Flow Cytometry | |
EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 | |
Human TMPRSS2 synthetic peptide, Accession # O15393 | |
0.1 mL | |
Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites | |
7113 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction