Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMPRSS2 Antibody (SR1968), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32216850UL
Description
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
TMPRSS2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR1968 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:60, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 | |
A synthesized peptide derived from human TMPRSS2 (Uniprot #: O15393) | |
50 μg | |
Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites | |
7113 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction