Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMPRSS2 Monoclonal Antibody (C1), Invitrogen™

Catalog No. FEMA551110
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551110 20 μL
FEMA551109 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551110 Supplier Invitrogen™ Supplier No. MA551110
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMPRSS2 is a protein that belongs to the serine protease family. The protein contains a type II transmembrane domain, a receptor class A domain, a scavenger receptor cysteine-rich domain and a protease domain. Serine proteases are known to be involved in many physiological and pathological processes. Its gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue. The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS2
Applications Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene TMPRSS2
Gene Accession No. O15393
Gene Alias D16Ertd61e; EC 3.4.219; epitheliasin; FLJ41954; MGC6821; Plasmic transmembrane protein X; PP9284; PRSS10; Serine protease 10; Tmprss2; TMPS2; Transmembrane protease serine 2; Transmembrane protease serine 2 catalytic chain; Transmembrane protease serine 2 non-catalytic chain; transmembrane protease, serine 2
Gene Symbols TMPRSS2
Host Species Mouse
Immunogen Recombinant Human TMPRSS2 protein, Ser284-Gly492 (Accession #O15393).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7113
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.