Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMPRSS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5141088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5141088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5141088 Supplier Invitrogen™ Supplier No. PA5141088
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMPRSS2 Polyclonal Antibody for Western Blot, IHC (P)

TMPRSS2 is a protein that belongs to the serine protease family. The protein contains a type II transmembrane domain, a receptor class A domain, a scavenger receptor cysteine-rich domain and a protease domain. Serine proteases are known to be involved in many physiological and pathological processes. Its gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue. The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TMPRSS2
Gene Accession No. O15393, Q9JIQ8
Gene Alias D16Ertd61e; EC 3.4.219; epitheliasin; FLJ41954; MGC6821; Plasmic transmembrane protein X; PP9284; PRSS10; Serine protease 10; Tmprss2; TMPS2; Transmembrane protease serine 2; Transmembrane protease serine 2 catalytic chain; Transmembrane protease serine 2 non-catalytic chain; transmembrane protease, serine 2
Gene Symbols TMPRSS2
Host Species Rabbit
Immunogen Synthetic peptide within human TMPRSS2 aa 300-350.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 156435, 50528, 7113
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.