Learn More
Invitrogen™ TMPRSS3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total TMPRSS3 protein.
Specifications
Specifications
| Antigen | TMPRSS3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Tmprss3 |
| Gene Accession No. | P57727, Q8K1T0 |
| Gene Alias | DFNB10; DFNB8; ECHOS1; Serine protease TADG-12; TADG12; TMPRSS3; Transmembrane protease serine 3; transmembrane protease, serine 3; transmembrane proteinase serine 3; transmembrane serine protease 3; tumor-associated differentially-expressed gene 12 protein; UNQ323/PRO382 |
| Gene Symbols | Tmprss3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.