Learn More
Invitrogen™ TMPRSS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534273
Description
Percent identity with other species by BLAST analysis: Human (100%); Chimpanzee, Orangutan, Gibbon (94%); Gorilla, Monkey (89%); Marmoset, Dog (83%). Antigen retrieval with HIER and sodium citrate at pH 6.0 is required for Immunohistochemistry using formalin-fixed paraffin-embedded human tissues.
This gene encodes a member of the serine protease family. Serine proteases are known to be involved in a variety of biological processes, whose malfunction often leads to human diseases and disorders. This gene was identified as a gene overexpressed in pancreatic carcinoma. The encoded protein is membrane bound with a N-terminal anchor sequence and a glycosylated extracellular region containing the serine protease domain. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
TMPRSS4 | |
Polyclonal | |
Unconjugated | |
TMPRSS4 | |
Cap2; CAPH2; Channel-activating protease 2; mCAP2; Membrane-type serine protease 2; MT-SP2; TMPRSS3; Tmprss4; Transmembrane protease serine 4; transmembrane protease, serine 4; transmembrane serine protease 3; type II membrane serine protease; UNQ776/PRO1570 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56649 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NRS4 | |
TMPRSS4 | |
Synthetic 18 amino acid peptide from N-Terminus of human TMPRSS4. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.