Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987180
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7180 Supplier Bioss Supplier No. BS6286R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the serine protease family. Serine proteases are known to be involved in a variety of biological processes, whose malfunction often leads to human diseases and disorders. This gene was identified as a gene overexpressed in pancreatic carcinoma. The encoded protein is membrane bound with a N-terminal anchor sequence and a glycosylated extracellular region containing the serine protease domain. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen TMPRSS4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TMPRSS4
Gene Accession No. Q9NRS4
Gene Alias Cap2; CAPH2; Channel-activating protease 2; mCAP2; Membrane-type serine protease 2; MT-SP2; TMPRSS3; Tmprss4; Transmembrane protease serine 4; transmembrane protease, serine 4; transmembrane serine protease 3; type II membrane serine protease; UNQ776/PRO1570
Gene Symbols TMPRSS4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TMPRSS4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56649
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.