Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS5 Monoclonal Antibody (OTI6E11), TrueMAB™, OriGene
SDP

Catalog No. 502540914
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0914 30 μL
50-167-4329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0914 Supplier OriGene Supplier No. TA503427S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMPRSS5 Monoclonal antibody specifically detects TMPRSS5 in Canine, Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen TMPRSS5
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI6E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pgam2
Gene Accession No. O70250, P15259
Gene Alias adrenal mitochondrial protease; Amp; MGC141886; MGC148044; Spinesin; Tmprss5; Transmembrane protease serine 5; transmembrane protease, serine 5; transmembrane protease, serine 5 (spinesin)
Host Species Mouse
Immunogen Full length human recombinant protein of human PGAM2 produced in HEK293 cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 475495, 5224, 56012
Target Species Canine, Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.