Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS5 Monoclonal Antibody (OTI6G10), TrueMAB™, OriGene
SDP

Catalog No. 502540913
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0913 30 μL
50-167-4330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0913 Supplier OriGene Supplier No. TA503429S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMPRSS5 Monoclonal antibody specifically detects TMPRSS5 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen TMPRSS5
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI6G10
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TMPRSS5
Gene Accession No. Q9H3S3
Gene Alias adrenal mitochondrial protease; Amp; MGC141886; MGC148044; Spinesin; Tmprss5; Transmembrane protease serine 5; transmembrane protease, serine 5; transmembrane protease, serine 5 (spinesin)
Host Species Mouse
Immunogen Full length human recombinant protein of human TMPRSS5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80975
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.