Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TMS(PEG)12 Branched Methyl-PEG-NHS-Ester Reagent

Catalog No. PI22424
Click to view available options
Quantity:
1 g

A three-branched, polyethylene glycol compound (3 times 12 PEG units) activated as an NHS ester for covalent PEGylation of primary amines on proteins or assay surfaces.

Thermo Scientific™ Pierce TMS(PEG)12 is a three-branched, polyethylene glycol compound (3 times 12 PEG units) activated as an NHS ester for covalent pegylation of primary amines (e.g., lysines) on proteins or assay surfaces.

TMS(PEG)12 is the abbreviation for a branched trimethyl (TM) and succinimide ester (S) derivative of polyethylene glycol (PEG) for efficient and specific modification of primary amines. Each methyl-terminated PEG (mPEG) branch contains 12 ethylene glycol units. The three branches are attached to a 4-unit PEG stem that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at the distal end. The NHS ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.

Highlights:

  • NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
  • Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Easy-to-follow instructions increase the likelihood of a successful outcome
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine
Content And Storage Store desiccated at -20°C.
Label Type Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer 12 ethylene glycol units
Form Solid
Quantity 1 g
Solubility Water
Product Type NHS-Ester Reagent
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.