Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMSB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550533 Supplier Invitrogen™ Supplier No. PA550533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total Thymosin beta 10 protein.

This protein plays an important role in the organization of the cytoskeleton. It binds to and sequesters actin monomers (G actin) and therefore inhibits actin polymerization. Experiment indicate that the expression decrease dramatically after birth; It expresses high levels in human fetal brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMSB10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TMSB10
Gene Accession No. P63312, P63313, Q6ZWY8
Gene Alias MIG12; migration-inducing gene 12; migration-inducing protein 12; prothymosin beta 10; Ptmb10; Tb10; Thyb10; thymosin; thymosin beta 10; thymosin beta-10; thymosin, beta 10; TMSB10
Gene Symbols TMSB10
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human thymosin beta 10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19240, 50665, 9168
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.