Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TMT™ Elution Buffer and Anti-TMT Resin

Catalog No. pi90104
Change view
Click to view available options
Quantity:
6 mL
20 mL
Product Type:
Immobilized Anti-TMT™ Antibody Resin
TMT™ Elution Buffer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PI90104 20 mL TMT™ Elution Buffer
PI90076 6 mL Immobilized Anti-TMT™ Antibody Resin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI90104 Supplier Thermo Scientific™ Supplier No. 90104
Only null left
Add to Cart
Edge
Add to Cart

For enhanced enrichment, use Thermo Scientific TMT Elution Buffer with Immobilized Anti-TMT Resin for peptides that have been chemically labeled with Tandem Mass Tag (TMT) Isobaric Label Reagents for multiplexed protein quantitation using tandem mass spectrometry.

Tandem mass tag (TMT) isobaric label reagents are used for multiplexed protein quantitation using tandem mass spectrometry. Immobilized Anti-TMT Resin is designed to effectively capture and enrich peptides or proteins labeled using any TMT reagent. Anti-TMT Resin and TMT Elution Buffer are effective for reducing sample complexity, improving dynamic range, and studying cysteine modifications such as S-nitrosylation, oxidation, and disulfide bonds of protein samples labeled with iodoTMT reagents.

Tandem mass tag (TMT) reagents are used to chemically label proteins in different samples for multiplexed quantitation using tandem mass spectrometry. Immobilized Anti-TMT Resin contains an agarose-immobilized anti-TMT monoclonal antibody and is effective for capture and enrichment of peptides or proteins labeled using any TMT reagent. This helps simplify otherwise complex samples before LC-MS/MS analysis, especially when the targets for labeling are rare (e.g., cysteine sulfhydryls). Enrichment of labeled peptides by this method results in fewer peptides identified per protein, but can significantly increase the number of unique proteins identified, making it easier to detect and measure low-abundance proteins.

Immobilized Anti-TMT Resin uses our highly specific Anti-TMT Antibody (Cat. No. 90075) to bind peptides labeled with TMT reagents. The antibody is specific for the mass reporter region of the TMT reagents, which allows for enrichment of TMT-labeled peptides regardless of peptide labeling chemistry (e.g., amine-reactive, sulfhydryl-reactive). TMT Elution Buffer is a volatile neutral buffer that competitively elutes captured TMT-labeled peptides. When used in combination with Immobilized Anti-TMT Resin, TMT Elution Buffer provides higher specific enrichment of TMT-labeled peptides compared to acidic or basic elution buffers.

Enrichment features include:
Specific—uses specific, high-affinity antibody for the TMT structure (Kd >100 pM)
Compatible—effective for all varieties of TMT reagents (amine-reactive or cysteine-reactive)
Versatile—use resin with TMT Elution Buffer for optimal sample enrichment

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type TMT™ Elution Buffer
Product Line TMT
Content And Storage Store at 4°C.
Label or Dye Tandem Mass Tag™ (TMT) Labels
Starting Material Cell Lysate
Final Product Type Peptide
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 20 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.