Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TMT11-131C Label Reagent

Catalog No. p-7180389
Click to view available options
Quantity:
1 x 5 mg
1 x 5 mg (per tag)
3 x 0.8 mg
3 x 0.8 mg (per tag)
8 x 40 μg​ (per tag), 80 reactions

Tandem mass tag (TMT) reagents are specially designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry.

Tandem mass tag (TMT) reagents are specially designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry. The TMT11-131C tag structure is identical to the other TMT10pex tags, but the TMT11-131C mass reporter is labeled with only 13C isotopes. The addition of the TMT11-131C tag enables separation of the 131-mass reporter ion of the original TMT10plex reagents into N and C variants, creating an 11th channel for relative quantitation using high resolution Orbitrap MS instruments.

The advantages of TMT label reagents include increased multiplexed relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. TMT10plex label reagents plus TMT11-131C is ideal for analysis of multiple protein samples from inhibitor dose response experiments, time course experiments, or to improve statistics on biological replicates.

Features of this TMT11-131C Label Reagent include:
Multiplex—concurrent MS analysis of up to 11 samples derived from cells, tissues, or biological fluids
Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
Efficient—amine-reactive, NHS-ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
Compatible—optimized for use with high resolution Thermo Scientific Orbirtrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer 2.3

TMT11-131C is available as a standalone tag (Cat. No. A34807, A37724) or as part of this TMT10plex Isobaric Label Reagent Set plus TMT11-131C. When combined with the industry-leading, high resolution Orbitrap instruments and software, TMT reagents provide integrated total solutions for quantitative protein expression analysis.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Isobaric Label Reagent
Content And Storage 1 vial, store at –5 to –30°C.
Label or Dye Tandem Mass Tag (TMT) Label
Shipping Condition Dry Ice
Starting Material Peptides
Detection Method Mass Spectrometry
Workflow Step Peptide labeling and quantitation
Quantity 1 x 5 mg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.