Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMTC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113094
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113094 Supplier Invitrogen™ Supplier No. PA5113094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMTC1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMTC1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Tmtc1
Gene Accession No. Q3UV71, Q8IUR5
Gene Alias Arg99; ARG99 homolog; BC023818; OLF; Protein O-mannosyl-transferase TMTC1; Tmtc1; TMTC1A; transmembrane and tetratricopeptide repeat containing 1; transmembrane and tetratricopeptide repeat containing 1A; transmembrane and TPR repeat-containing protein 1
Gene Symbols Tmtc1
Host Species Rabbit
Immunogen Recombinant Human Transmembrane and TPR repeat-containing protein 1 protein (1-227AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387314, 83857
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.