Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMTC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP185027
Description
TMTC2 Polyclonal specifically detects TMTC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TMTC2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
DKFZp762A217, transmembrane and tetratricopeptide repeat containing 2, transmembrane and TPR repeat-containing protein 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
160335 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TMTC2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MMGEAYMRLSKLPEAEHWYMESLRSKTDHIPAHLTYGKLLALTGRKSEAEKLFLKAIELDPTKGNCYMHYGQFLLEEARLIEAAEMAKKAAELDSTEFDVVFNAAHMLRQASLNEAAEKYYDLAARLRPNYPAALMNLGAIL | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction