Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMTC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569360
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
This gene encodes a transmembrane protein that belongs to family of proteins containing an N-terminal transmembrane domain and a C-terminal tetratricopeptide repeat (TPR) domain. TPR domains mediate protein-protein interactions in various cellular processes, such as synaptic vesicle fusion, protein folding, and protein translocation. A pseudogene of this gene has been defined on chromosome 5.
Specifications
TMTC4 | |
Polyclonal | |
Unconjugated | |
TMTC4 | |
4930403J22Rik; 5730419O14Rik; Protein O-mannosyl-transferase TMTC4; protein O-mannosyl-transferase TMTC4; transmembrane and TPR repeat-containing protein 4; RGD1560183; si:ch211-168k14.1; Tmtc4; transmembrane and tetratricopeptide repeat containing 4; transmembrane and TPR repeat-containing protein 4; transmembrane and TPR repeat-containing protein 4-like protein; transmembrane O-mannosyltransferase targeting cadherins 4; wu:fc58d04 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84899 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5T4D3 | |
TMTC4 | |
synthetic peptide directed towards the N terminal region of human TMTC4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction