Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMTC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553432 Supplier Invitrogen™ Supplier No. PA553432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLGRLYADLN RHVDALNAWR NATVLKPEHS LAWNNMIILL DNTGNLAQAE AVGREALELI PNDHSLMFSL ANVLGKSQKY KESEALFLKA IKANPNAASY HGNLAVLYHR WGHLDLAKKH YEISLQLDPT ASGTKENYGL LR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

This gene encodes a transmembrane protein that belongs to family of proteins containing an N-terminal transmembrane domain and a C-terminal tetratricopeptide repeat (TPR) domain. TPR domains mediate protein-protein interactions in various cellular processes, such as synaptic vesicle fusion, protein folding, and protein translocation. A pseudogene of this gene has been defined on chromosome 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMTC4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMTC4
Gene Accession No. Q5T4D3
Gene Alias 4930403J22Rik; 5730419O14Rik; Protein O-mannosyl-transferase TMTC4; protein O-mannosyl-transferase TMTC4; transmembrane and TPR repeat-containing protein 4; RGD1560183; si:ch211-168k14.1; Tmtc4; transmembrane and tetratricopeptide repeat containing 4; transmembrane and TPR repeat-containing protein 4; transmembrane and TPR repeat-containing protein 4-like protein; transmembrane O-mannosyltransferase targeting cadherins 4; wu:fc58d04
Gene Symbols TMTC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMTC4. Recombinant protein control fragment (Product #RP-91262).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.