Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMUB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556143 Supplier Invitrogen™ Supplier No. PA556143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATDSMRGEAP GAETPSLRHR GQAAQPEPST GFTATPPAPD SPQEPLVLRL KFLNDSEQVA RAWPHDTIGS LKRTQFPGRE QQVRLIYQGQ LLGDDTQTLG SLHLPPNCVL HC Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

TMUB1 gene ontology annotations related to this gene include ubiquitin-dependent ERAD pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMUB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMUB1
Gene Accession No. Q9BVT8
Gene Alias 2010004O20Rik; AB030183; C7orf21; Dendritic cell-derived ubiquitin-like protein; DULP; Hepatocyte odd protein shuttling protein; Hops; iHOPS; SB144; TMUB1; transmembrane and ubiquitin like domain containing 1; transmembrane and ubiquitin-like domain containing 1; transmembrane and ubiquitin-like domain-containing protein 1; ubiquitin-like protein DULP; ubiquitin-like protein SB144; UNQ763/PRO1555
Gene Symbols TMUB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMUB1. Recombinant protein control fragment (Product #RP-91092).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83590
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.