Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMUB2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300175F
Description
TMUB2 Polyclonal specifically detects TMUB2 in Human samples. It is validated for ELISA.Specifications
TMUB2 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
79089 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
MGC3123, transmembrane and ubiquitin-like domain containing 2, transmembrane and ubiquitin-like domain-containing protein 2 | |
Produced in rabbits immunized with purified, recombinant TMUB2 (NP_803190.2; Tyr37-Ser245). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction