Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMUB2 Monoclonal Antibody (09)
GREENER_CHOICE

Catalog No. PIMA530939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530939 Supplier Invitrogen™ Supplier No. MA530939
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TMUB2.

TMUB2 (Transmembrane And Ubiquitin Like Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is TMUB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMUB2
Applications ELISA
Classification Monoclonal
Clone 09
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TMUB2
Gene Accession No. Q71RG4
Gene Alias 2010008E23Rik; FP2653; Tmub2; transmembrane and ubiquitin like domain containing 2; transmembrane and ubiquitin-like domain containing 2; transmembrane and ubiquitin-like domain-containing protein 2; UNQ1897/PRO4343
Gene Symbols TMUB2
Host Species Mouse
Immunogen Recombinant Human TMUB2 Protein (Tyr37-Ser245).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79089
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.