Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMX Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA517954
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
TXNDC1 is a thioredoxin (TXN; see MIM 187700)-related protein with disulfide reductase activity (Matsuo et al., 2001 [PubMed 11152479]).
Specifications
TMX | |
Polyclonal | |
Unconjugated | |
TMX1 | |
2810425A04Rik; AI465319; DKFZP564E1962; PDIA11; protein disulfide isomerase family A, member 11; PSEC0085; thioredoxin domain containing 1; thioredoxin domain-containing protein 1; thioredoxin related transmembrane protein 1; thioredoxin-related transmembrane protein 1; TMX; Tmx1; Transmembrane Trx-related protein; TXNDC; TXNDC1; UNQ235/PRO268 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
81542 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9H3N1 | |
TMX1 | |
Synthetic peptide sequence (RSLGPSLATDKS) corresponding to the C-terminus amino acids of TMX1 (aa 269-280). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction