Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109348 Supplier Invitrogen™ Supplier No. PA5109348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Daudi cell lysates, MG-63, human skin tissue, human breast carcinoma tissue, F9.

TXNDC1 is a thioredoxin (TXN; see MIM 187700)-related protein with disulfide reductase activity (Matsuo et al., 2001 [PubMed 11152479]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMX
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene TMX1
Gene Accession No. Q9H3N1
Gene Alias 2810425A04Rik; AI465319; DKFZP564E1962; PDIA11; protein disulfide isomerase family A, member 11; PSEC0085; thioredoxin domain containing 1; thioredoxin domain-containing protein 1; thioredoxin related transmembrane protein 1; thioredoxin-related transmembrane protein 1; TMX; Tmx1; Transmembrane Trx-related protein; TXNDC; TXNDC1; UNQ235/PRO268
Gene Symbols TMX1
Host Species Rabbit
Immunogen Synthetic peptide within Human TMX1 aa 89-138.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81542
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.