Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA133100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA133100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA133100 Supplier Invitrogen™ Supplier No. PA133100
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX74441-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TXNDC1 is a thioredoxin (TXN; see MIM 187700)-related protein with disulfide reductase activity (Matsuo et al., 2001 [PubMed 11152479]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TMX1
Gene Accession No. Q9H3N1
Gene Alias 2810425A04Rik; AI465319; DKFZP564E1962; PDIA11; protein disulfide isomerase family A, member 11; PSEC0085; thioredoxin domain containing 1; thioredoxin domain-containing protein 1; thioredoxin related transmembrane protein 1; thioredoxin-related transmembrane protein 1; TMX; Tmx1; Transmembrane Trx-related protein; TXNDC; TXNDC1; UNQ235/PRO268
Gene Symbols TMX1
Host Species Goat
Immunogen Peptide with sequence C-RSLGPSLATDKS, from the C Terminus.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81542
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.