Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMX1 Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
SDP

Catalog No. 501676871
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-6871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-6871 Supplier OriGene Supplier No. TA506948
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TXNDC1 is a thioredoxin -related protein with disulfide reductase activity.

Specifications

Antigen TMX1
Applications Western Blot
Classification Monoclonal
Clone OTI1D8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TMX1
Gene Accession No. Q9H3N1
Gene Alias PDIA11, TMX, TXNDC, TXNDC1
Gene Symbols TMX1
Host Species Mouse
Immunogen Full length human recombit protein of human TMX1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81542
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.