Learn More
TMX1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304040A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
TXNDC1 is a thioredoxin -related protein with disulfide reductase activity .Specifications
TMX1 | |
Polyclonal | |
Unconjugated | |
TMX1 | |
DKFZp564E1962, PDIA11, TMX, TXNDC, TXNDC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81542 | |
4° C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9H3N1 | |
TMX1 | |
Between 230 and 280 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.