Learn More
TMX1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TMX1 |
---|---|
Concentration | 767 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8568
![]() ![]() |
Proteintech Group Inc
274891AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The TMX1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TMX1. This antibody recognizes human, mouse, rat antigen. The TMX1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TMX1 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
767 μg/mL | |
Polyclonal | |
Rabbit | |
81542 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.