Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMX2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP288444
Description
TMX2 Polyclonal specifically detects TMX2 in Mouse samples. It is validated for Western Blot.Specifications
TMX2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
51075 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Cell proliferation-inducing gene 26 protein, DKFZp781O2021, growth-inhibiting gene 11, MGC111151, PDIA12, PIG26, protein disulfide isomerase family A, member 12, thioredoxin domain containing 14, Thioredoxin domain-containing protein 14, thioredoxin-related transmembrane protein 2, TXNDC14 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse TMX2 | |
100 μL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction