Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564133
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564133 Supplier Invitrogen™ Supplier No. PA564133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ENVIREFNLN ELYQRAKKLS KAGDNIPEEQ PVASTPTTVS DGENKKDK Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.

This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and a C-terminal ER-retention sequence. This protein is enriched on the mitochondria-associated-membrane of the ER via palmitoylation of two of its cytosolically exposed cysteines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMX2
Gene Accession No. Q9Y320
Gene Alias 2310042M24Rik; AA589631; Cell proliferation-inducing gene 26 protein; CGI-31; growth-inhibiting gene 11; My009; PDIA12; PIG26; protein disulfide isomerase family A, member 12; PSEC0045; thioredoxin domain containing 14; thioredoxin domain-containing protein 14; thioredoxin related transmembrane protein 2; thioredoxin-related transmembrane protein 2; thioredoxin-related transmembrane protein-like protein; TMX2; TXNDC14; Unknown (protein for MGC:134152); UNQ237/PRO270
Gene Symbols TMX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMX2. Recombinant protein control fragment (Product #RP-104197).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.