Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569450
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and a C-terminal ER-retention sequence. This protein is enriched on the mitochondria-associated-membrane of the ER via palmitoylation of two of its cytosolically exposed cysteines.
Specifications
TMX2 | |
Polyclonal | |
Unconjugated | |
TMX2 | |
2310042M24Rik; AA589631; Cell proliferation-inducing gene 26 protein; CGI-31; growth-inhibiting gene 11; My009; PDIA12; PIG26; protein disulfide isomerase family A, member 12; PSEC0045; thioredoxin domain containing 14; thioredoxin domain-containing protein 14; thioredoxin related transmembrane protein 2; thioredoxin-related transmembrane protein 2; thioredoxin-related transmembrane protein-like protein; TMX2; TXNDC14; Unknown (protein for MGC:134152); UNQ237/PRO270 | |
Rabbit | |
Affinity chromatography | |
RUO | |
66958 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9D710 | |
TMX2 | |
synthetic peptide directed towards the C-terminal region of mouse TMX2. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction