Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMX4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169606
Description
TMX4 Polyclonal specifically detects TMX4 in Human samples. It is validated for Western Blot.Specifications
TMX4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DJ971N18.2, KIAA1162Thioredoxin domain-containing protein 13, PDIA14, protein disulfide isomerase family A, member 14, thioredoxin domain containing 13, thioredoxin-related transmembrane protein 4, TXNDC13 | |
Rabbit | |
39 kDa | |
100 μL | |
Primary | |
Canine: 86%; Porcine: 86%; Guinea pig: 85%; Mouse: 79%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9H1E5 | |
TMX4 | |
Synthetic peptides corresponding to TXNDC13(thioredoxin domain containing 13) The peptide sequence was selected from the C terminal of TXNDC13. Peptide sequence PGEDGVTREEVEPEEAEEGISEQPCPADTEVVEDSLRQRKSQHADKGL. | |
Affinity purified | |
RUO | |
56255 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction