Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119745 Supplier Invitrogen™ Supplier No. PA5119745
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Rat cerebellum tissue lysates, rat skin tissue lysates, human skin tissue, mouse testis tissue. Subcellular Location: Nucleus inner membrane.

This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and C-terminal ASP/GLU-rich calcium binding domain. Unlike most members of this gene family, it lacks a C-terminal ER-retention sequence. The encoded protein has been shown to have reductase activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMX4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TMX4
Gene Accession No. Q8C0L0, Q9H1E5
Gene Alias 2810417D04Rik; 4930500L08Rik; AI843224; AW046784; D2Bwg1356e; DJ971N18.2; Kiaa1162; mKIAA1162; PDIA14; protein disulfide isomerase family A, member 14; PSEC0095; RGD1305146; thioredoxin domain containing 13; thioredoxin domain-containing protein 13; thioredoxin related transmembrane protein 4; thioredoxin-related transmembrane protein 4; Tmx4; Txndc13; UNQ475/PRO938
Gene Symbols TMX4
Host Species Rabbit
Immunogen Recombinant protein within human TMX4 aa 1-180.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296182, 52837, 56255
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.