Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tn mIgM (STn-PS A1), clone KT-IgM-8 Mouse anti-Human, Unconjugated, Clone: Kt-IgM-8, MilliporeSigma™

Catalog No. MABC17171MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC17171MI 100 μL
MABC17172MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC17171MI Supplier MilliporeSigma Supplier No. MABC1717100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tn mIgM (STn-PS A1), clone KT-IgM-8 Monoclonal antibody specifically detects Tn mIgM (STn-PS A1), clone KT-IgM-8 in Human samples. It is validated for ELISA, Flow Cytometry, Function Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tn mIgM (STn-PS A1), clone KT-IgM-8
Applications ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone Kt-IgM-8
Conjugate Unconjugated
Gene Alias Thomsen-nouveau (Tn);Tn antigen;Tn saccharide;N-acetyl Galactosamine;Alpha-GalNAc;Alpha GalpNAc;GalNAc;GalNAc-alpha;N-acetyl-alpha-D-galactosamine;Blood group A antigen
Host Species Mouse
Immunogen Semi-synthetic Tn-PS A1 (Tn= D-GalpNAc)
Purification Method Ligatrap
Quantity 100 μL
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone KT-IgM-8 is a mouse monoclonal antibody that detects Thomsen-nouveau (Tn) polysaccharide A1 monovalently, including in human tumor cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.