Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha Antibody (6402) [Allophycocyanin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10564A
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
TNF-alpha | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
6402 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
E. coli-derived recombinant human TNF-alpha , Gly57-Leu233, Accession # P01375 | |
0.1 mL | |
Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
7124 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction