Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUCNR1/GPR91 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 14 publications
$247.53 - $591.43
Specifications
| Antigen | SUCNR1/GPR91 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, In vitro assay reported in scientific literature (PMID 30478422) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP100861SS
![]() |
Novus Biologicals
NBP100861SS |
0.025 mg |
Each for $247.53
|
|
|||||
NBP100861
![]() |
Novus Biologicals
NBP100861 |
0.1 mg |
Each for $591.43
|
|
|||||
Description
SUCNR1/GPR91 Polyclonal specifically detects SUCNR1/GPR91 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In vitro assay.Specifications
| SUCNR1/GPR91 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, In vitro assay reported in scientific literature (PMID 30478422) | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9BXA5 | |
| 56670 | |
| A synthetic peptide made to an internal portion of the human GPR91 protein (between residues 100-200) [Uniprot: Q9BXA5] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 38 kDa |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| GPR91G protein-coupled receptor 91, G-protein coupled receptor 91, P2Y purinoceptor 1, P2Y purinoceptor 1-like, succinate receptor 1 | |
| SUCNR1 | |
| IgG | |
| Affinity Purified | |
| GPR91 (A135) detects endogenous levels of GPR91 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title